Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001001321.3 | NP_001001321.2 | 4 | 307 | |
GenBank mRNA | AB117931.1 | - | 4 | 288 | |
AB117932.1 | - | 4 | 288 | ||
AK154257.1 | - | 4 | 333 | ||
AK159148.1 | - | 4 | 308 | ||
BC107210.1 | - | 3 | 254 | ||
BC107211.1 | - | 4 | 287 | 40051417 |