Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010706.2 | NP_034836.1 | 5 | 880 | |
GenBank mRNA | AF026795.1 | - | 4 | 430 | |
AF510729.1 | - | 5 | 529 | ||
AK145734.1 | - | 5 | 700 | ||
AY044870.1 | - | 5 | 552 | ||
BC011236.1 | - | 5 | 518 | 4205669 | |
BC021632.1 | - | 5 | 536 | 4218531 | |
BC030297.1 | - | 5 | 693 | 4986324 | |
BC094008.1 | - | 5 | 531 | 4986911 | |
BC141106.1 | - | 5 | 602 | 9055718 | |
BC145387.1 | - | 5 | 602 | 9053945 | |
EF017938.1 | - | 5 | 880 | ||
EF017939.1 | - | 5 | 880 |