Gene Transcripts

Gene Symbol
Ypel5
Entrez Gene ID
383295
Gene Name
yippee-like 5 (Drosophila)
Gene Aliases
2310076K21Rik, CGI-127
Chromosome Location
Chr. 17: 72836625 - 72851195 on Build GRCm38
UniGene ID
Mm.277533
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027166.5 NP_081442.1 3 598
GenBank mRNA AK046281.1 - 3 597
AK049096.1 - 3 599
AK049644.1 - 3 597
AK137446.1 - 3 570
AK154461.1 - 3 637
AK165943.1 - 3 600
AK167998.1 - 3 614
AK169241.1 - 3 601
AK169986.1 - 3 572
AK171010.1 - 3 599
BC025125.1 - 1 359 4921751
BC085109.1 - 3 617 30737658

View Full Product Details