Gene Transcripts
- Gene Symbol
- Krt72
- Entrez Gene ID
- 105866
- Gene Name
- keratin 72
- Gene Aliases
- AI507495, K6irs2, Krt2-35, Krt72-ps
- Chromosome Location
- Chr. 15: 101776560 - 101786458 on Build GRCm38
- UniGene ID
- Mm.298210
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_213728.1 |
NP_998893.1
|
4 |
782 |
|
View Full Product Details