Gene Transcripts

Gene Symbol
Fam19a1
Entrez Gene ID
320265
Gene Name
family with sequence similarity 19, member A1
Gene Aliases
C630007B19Rik, FAM19A1 TAFA-1, TAFA1, Tafa-1
Chromosome Location
Chr. 6: 96112775 - 96657204 on Build GRCm38
UniGene ID
Mm.489890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_182808.3 NP_877960.1 5 2188
GenBank mRNA AK049880.1 - 4 2079
AK050001.1 - 5 2190

View Full Product Details