Gene Transcripts

Gene Symbol
Ly6g5c
Entrez Gene ID
294245
Gene Name
lymphocyte antigen 6 complex, locus G5C
Gene Aliases
re8
Chromosome Location
Chr. 20: 5100415 - 5104494 on Build Rnor_6.0
UniGene ID
Rn.213816
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198739.3 NP_942034.3 2 279
XM_008772716.1 XP_008770938.1 2 280
GenBank mRNA AJ515240.1 - 2 279

View Full Product Details