Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001301015.1 | NP_001287944.1 | 23 | 4054 | |
NM_001301018.1 | NP_001287947.1 | 22 | 3429 | ||
NM_001301027.1 | NP_001287956.1 | 20 | 3800 | ||
NM_001301028.1 | NP_001287957.1 | 19 | 2466 | ||
NM_014945.3 | NP_055760.1 | 24 | 3700 | ||
XM_005268392.1 | XP_005268449.1 | 23 | 3582 | ||
XM_005268393.1 | XP_005268450.1 | 22 | 3534 | ||
XM_005268394.1 | XP_005268451.1 | 22 | 3495 | ||
XM_005268395.1 | XP_005268452.1 | 21 | 3396 | ||
XM_005268396.1 | XP_005268453.1 | 20 | 3348 | ||
XM_017009238.1 | XP_016864727.1 | 20 | 3490 | ||
GenBank mRNA | AB020650.1 | - | 24 | 3613 | |
AK098469.1 | - | 1 | 2004 | ||
AK225769.1 | - | 23 | 3922 | ||
AL833021.1 | - | 17 | 2808 | ||
BC001665.2 | - | 19 | 2128 | 2988043 |