Gene Transcripts

Gene Symbol
ZC3H13
Entrez Gene ID
23091
Gene Name
zinc finger CCCH-type containing 13
Gene Aliases
KIAA0853
Chromosome Location
Chr. 13: 45954469 - 46052785 on Build GRCh38
UniGene ID
Hs.136102
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015070.4 NP_055885.3 17 5241
XM_005266312.1 XP_005266369.1 17 5535
GenBank mRNA AL136745.1 - 10 4277
AY283618.1 - 17 5239

View Full Product Details