Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286715.1 | NP_001273644.1 | 16 | 2519 | |
NM_001286717.1 | NP_001273646.1 | 16 | 2399 | ||
NM_015239.2 | NP_056054.2 | 17 | 2464 | ||
XM_005251847.3 | XP_005251904.1 | 17 | 2530 | ||
XM_005251848.2 | XP_005251905.1 | 17 | 2708 | ||
XM_005251849.3 | XP_005251906.1 | 16 | 2454 | ||
XM_011518418.2 | XP_011516720.1 | 17 | 2530 | ||
XM_011518420.2 | XP_011516722.1 | 7 | 5783 | ||
XM_017014544.1 | XP_016870033.1 | 17 | 3075 | ||
XM_017014545.1 | XP_016870034.1 | 17 | 2410 | ||
XM_017014546.1 | XP_016870035.1 | 16 | 2297 | ||
XM_017014547.1 | XP_016870036.1 | 16 | 2454 | ||
XM_017014548.1 | XP_016870037.1 | 16 | 2397 | ||
XM_017014549.1 | XP_016870038.1 | 15 | 2416 | ||
GenBank mRNA | AB028958.2 | - | 17 | 2442 | |
AK001544.1 | - | 18 | 2264 | ||
AK022562.1 | - | 5 | 1107 | ||
AK023280.1 | - | 5 | 1099 | ||
AK024698.1 | - | 1 | 49 | ||
AK295304.1 | - | 13 | 1958 | ||
AK295774.1 | - | 16 | 2399 | ||
AK299506.1 | - | 13 | 1959 | ||
AL833359.1 | - | 5 | 1056 | ||
BC060815.1 | - | 17 | 2429 | 30344122 | |
BX648366.1 | - | 5 | 1511 |