Gene Transcripts

Gene Symbol
SH2B1
Entrez Gene ID
25970
Gene Name
SH2B adaptor protein 1
Gene Aliases
PSM, SH2B
Chromosome Location
Chr. 16: 28846600 - 28874213 on Build GRCh38
UniGene ID
Hs.15744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145795.1 NP_001139267.1 4 1328
NM_001145796.1 NP_001139268.1 4 1328
NM_001145797.1 NP_001139269.1 4 1328
NM_001145812.1 NP_001139284.1 4 1427
NM_001308293.1 NP_001295222.1 6 1584
NM_001308294.1 NP_001295223.1 3 263
NM_015503.2 NP_056318.2 3 3158
XM_017023114.1 XP_016878603.1 3 1747
XM_017023115.1 XP_016878604.1 5 1454
XM_017023116.1 XP_016878605.1 4 2812
XM_017023117.1 XP_016878606.1 3 1747
XM_017023118.1 XP_016878607.1 3 434
XM_017023119.1 XP_016878608.1 3 263
XM_017023120.1 XP_016878609.1 2 74
GenBank mRNA AB037720.1 - 3 4347
AF227967.1 - 4 1184
AF227968.1 - 4 1184
AF227969.1 - 4 1184
AK027488.1 - 3 493
AK055104.1 - 6 1495
AK290332.1 - 4 1427
AK297080.1 - 3 214
AK298569.1 - 3 313
AL049924.1 - 3 628
AL713760.1 - 3 3158
BC010704.1 - 3 881 3866324
DB026545.1 - 3 263

View Full Product Details