Gene Transcripts

Gene Symbol
RNF19A
Entrez Gene ID
25897
Gene Name
ring finger protein 19A, RBR E3 ubiquitin protein ligase
Gene Aliases
RNF19
Chromosome Location
Chr. 8: 100257059 - 100336218 on Build GRCh38
UniGene ID
Hs.292882
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001280539.1 NP_001267468.1 3 785
NM_015435.4 NP_056250.3 3 816
NM_183419.3 NP_904355.1 2 892
XM_005250853.3 XP_005250910.1 2 854
XM_011516956.2 XP_011515258.1 2 877
XM_017013302.1 XP_016868791.1 3 977
XM_017013303.1 XP_016868792.1 3 887
XM_017013304.1 XP_016868793.1 2 834
XM_017013305.1 XP_016868794.1 1 581
GenBank mRNA AB029316.1 - 3 816
BC093938.1 - 1 588 7939783
BC093940.2 - 1 588 7939785

View Full Product Details