Gene Transcripts
- Gene Symbol
- CDC40
- Entrez Gene ID
- 51362
- Gene Name
- cell division cycle 40
- Gene Aliases
- EHB3, PRP17, PRPF17
- Chromosome Location
- Chr. 6: 110180421 - 110232220 on Build GRCh38
- UniGene ID
- Hs.428147
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_015891.2 |
NP_056975.1
|
1, 2 |
209 |
|
GenBank mRNA |
AB208903.1 |
- |
1, 2 |
183 |
|
AF015044.1 |
- |
1, 2 |
176 |
|
AF038392.1 |
- |
1, 2 |
209 |
|
AF061241.1 |
- |
1, 2 |
182 |
|
AK222721.1 |
- |
1, 2 |
206 |
|
AK295055.1 |
- |
1, 2 |
202 |
|
AK314601.1 |
- |
1, 2 |
205 |
|
BC111061.1 |
- |
2, 3 |
286 |
|
BC117145.1 |
- |
1, 2 |
206 |
40125696 |
BC126114.1 |
- |
1, 2 |
206 |
8991830 |
View Full Product Details