Gene Transcripts

Gene Symbol
KCNK12
Entrez Gene ID
56660
Gene Name
potassium two pore domain channel subfamily K member 12
Gene Aliases
K2p12.1, THIK-2, THIK2
Chromosome Location
Chr. 2: 47520775 - 47570331 on Build GRCh38
UniGene ID
Hs.591586
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022055.1 NP_071338.1 2 880
GenBank mRNA AF287302.1 - 2 880
BC047749.1 - 2 908 6186991

View Full Product Details