Gene Transcripts

Gene Symbol
SCCPDH
Entrez Gene ID
51097
Gene Name
saccharopine dehydrogenase (putative)
Gene Aliases
CGI-49, NET11
Chromosome Location
Chr. 1: 246724076 - 246768137 on Build GRCh38
UniGene ID
Hs.498397
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016002.2 NP_057086.2 6, 7 1050
GenBank mRNA AF151807.1 - 6, 7 700
AK075178.1 - 6, 7 806
BC026185.2 - 6, 7 711 4798194

View Full Product Details