Gene Transcripts

Gene Symbol
INSIG2
Entrez Gene ID
51141
Gene Name
insulin induced gene 2
Gene Aliases
INSIG-2
Chromosome Location
Chr. 2: 118088418 - 118110031 on Build GRCh38
UniGene ID
Hs.7089
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001321329.1 NP_001308258.1 2 357
NM_016133.3 NP_057217.2 2 364
GenBank mRNA AF125392.1 - 1 278
AF527632.1 - 2 313
AK291433.1 - 2 312
BC022475.1 - 2 308 4794938
HY053967.1 - 2 366

View Full Product Details