Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016131.4 | NP_057215.3 | 3 | 961 | |
GenBank mRNA | AF086917.1 | - | 3 | 306 | |
AF106681.1 | - | 3 | 302 | ||
AF297660.1 | - | 3 | 212 | ||
AF498945.1 | - | 3 | 212 | ||
AK023223.1 | - | 3 | 711 | ||
AL136650.1 | - | 3 | 720 | ||
AL517754.3 | - | 3 | 722 | ||
BC000896.1 | - | 3 | 287 | 3464547 | |
CR457303.1 | - | 3 | 212 |