Gene Transcripts
- Gene Symbol
- MRPS18C
- Entrez Gene ID
- 51023
- Gene Name
- mitochondrial ribosomal protein S18C
- Gene Aliases
- CGI-134, MRP-S18-1, MRP-S18-c, MRPS18-1, S18mt-c, mrps18-c
- Chromosome Location
- Chr. 4: 83455932 - 83461723 on Build GRCh38
- UniGene ID
- Hs.334772
- Species
- Homo sapiens
- Gene Symbol
- FAM175A
- Entrez Gene ID
- 84142
- Gene Name
- family with sequence similarity 175 member A
- Gene Aliases
- ABRA1, CCDC98
- Chromosome Location
- Chr. 4: 83459515 - 83485181 on Build GRCh38
- UniGene ID
- Hs.334772
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BQ775930.1 |
- |
1 |
664 |
|
View Full Product Details