Gene Transcripts
- Gene Symbol
- DCTN4
- Entrez Gene ID
- 51164
- Gene Name
- dynactin subunit 4
- Gene Aliases
- DYN4, P62
- Chromosome Location
- Chr. 5: 150708747 - 150759095 on Build GRCh38
- UniGene ID
- Hs.675564
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135643.1 |
NP_001129115.1
|
1, 2 |
245 |
|
NM_016221.3 |
NP_057305.1
|
1, 2 |
245 |
|
XM_005268457.1 |
XP_005268514.1
|
1, 2 |
183 |
|
XM_011537646.1 |
XP_011535948.1
|
1, 2 |
245 |
|
GenBank mRNA |
AF195120.1 |
- |
1, 2 |
160 |
|
AK000299.1 |
- |
1, 2 |
164 |
|
AK296787.1 |
- |
1, 2 |
164 |
|
AK307757.1 |
- |
1, 2 |
184 |
|
AK311693.1 |
- |
1, 2 |
181 |
|
BC026323.1 |
- |
1, 2 |
170 |
4815273 |
DC321382.1 |
- |
1, 2 |
245 |
|
KU178637.1 |
- |
1, 2 |
143 |
|
KU178638.1 |
- |
1, 2 |
143 |
|
View Full Product Details