Gene Transcripts

Gene Symbol
SNX9
Entrez Gene ID
51429
Gene Name
sorting nexin 9
Gene Aliases
SDP1, SH3PX1, SH3PXD3A, WISP
Chromosome Location
Chr. 6: 157716046 - 157945077 on Build GRCh38
UniGene ID
Hs.191213
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016224.4 NP_057308.1 2 345
XM_005267015.2 XP_005267072.1 4 423
GenBank mRNA AF001629.1 - 2 150
AF121859.1 - 2 105
AF131214.1 - 2 124
AF172847.1 - 2 225
AK314360.1 - 2 179
BC005022.1 - 2 252 3832234
BG766499.1 - 2 255
DA315945.1 - 2 346
DA336311.1 - 2 291

View Full Product Details