Gene Transcripts

Gene Symbol
HECA
Entrez Gene ID
51696
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
HDC, HDCL, HHDC, dJ225E12.1
Chromosome Location
Chr. 6: 139135112 - 139180809 on Build GRCh38
UniGene ID
Hs.197644
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016217.2 NP_057301.1 2 638
GenBank mRNA AB033492.1 - 2 638
BC070068.1 - 2 646 30340910

View Full Product Details