Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286682.1 | NP_001273611.1 | 4 | 463 | |
NM_001286683.1 | NP_001273612.1 | 4 | 536 | ||
NM_016228.3 | NP_057312.1 | 3 | 669 | ||
NM_182662.1 | NP_872603.1 | 4 | 451 | ||
XM_006714231.2 | XP_006714294.1 | 5 | 593 | ||
GenBank mRNA | AF481738.1 | - | 3 | 670 | |
AK055952.1 | - | 4 | 451 | ||
AK314729.1 | - | 4 | 538 | ||
BC031068.1 | - | 4 | 456 | 5286180 |