Gene Transcripts

Gene Symbol
R3HCC1L
Entrez Gene ID
27291
Gene Name
R3H domain and coiled-coil containing 1 like
Gene Aliases
C10orf28, GIDRP86, GIDRP88, PSORT
Chromosome Location
Chr. 10: 98134608 - 98244897 on Build GRCh38
UniGene ID
Hs.419800
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256619.1 NP_001243548.1 11 3282
NM_001256620.1 NP_001243549.1 9 3189
NM_001256621.1 NP_001243550.1 8 1386
NM_014472.4 NP_055287.4 9 3185
NM_138469.2 NP_612478.2 10 3282
XM_011539639.1 XP_011537941.1 11 3539
XM_011539640.1 XP_011537942.1 12 3344
XM_011539641.1 XP_011537943.1 11 3296
XM_011539642.1 XP_011537944.1 12 3636
XM_011539643.1 XP_011537945.1 10 3223
XM_011539644.1 XP_011537946.1 10 3227
XM_011539645.2 XP_011537947.1 10 3470
XM_011539646.1 XP_011537948.1 9 3102
XM_011539647.1 XP_011537949.1 9 3104
XM_011539648.1 XP_011537950.1 8 2977
XM_011539649.1 XP_011537951.1 10 3497
XM_017016074.1 XP_016871563.1 10 4435
XM_017016075.1 XP_016871564.1 7 2935
XM_017016076.1 XP_016871565.1 10 3229
XM_017016077.1 XP_016871566.1 12 3392
XM_017016078.1 XP_016871567.1 11 3314
XM_017016079.1 XP_016871568.1 11 3328
XM_017016080.1 XP_016871569.1 9 3234
XM_017016081.1 XP_016871570.1 10 3316
XM_017016082.1 XP_016871571.1 8 3108
XM_017016083.1 XP_016871572.1 9 3124
GenBank mRNA AF525304.1 - 11 3235
AK058187.1 - 5 976
AK094479.1 - 6 1696
BC009231.2 - 5 1111
BC047908.1 - 8 1332

View Full Product Details