Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136197.1 | NP_001129669.1 | 11 | 1409 | |
NM_001136198.1 | NP_001129670.1 | 13 | 1685 | ||
NM_016263.3 | NP_057347.2 | 14 | 1854 | ||
XM_005259573.4 | XP_005259630.1 | 14 | 2045 | ||
XM_017026863.1 | XP_016882352.1 | 15 | 2389 | ||
GenBank mRNA | AB033068.1 | - | 13 | 1676 | |
AF080397.1 | - | 13 | 1654 | ||
AF083810.1 | - | 14 | 1781 | ||
AF102508.1 | - | 4 | 591 | ||
BC013413.2 | - | 14 | 1831 | 3160334 | |
BC111760.1 | - | 1 | 156 |