Gene Transcripts
- Gene Symbol
- KLHL7
- Entrez Gene ID
- 55975
- Gene Name
- kelch like family member 7
- Gene Aliases
- KLHL6, SBBI26
- Chromosome Location
- Chr. 7: 23105734 - 23175421 on Build GRCh38
- UniGene ID
- Hs.654817
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001031710.2 |
NP_001026880.2
|
3, 4 |
610 |
|
NM_001172428.1 |
NP_001165899.1
|
3, 4 |
610 |
|
NM_018846.4 |
NP_061334.4
|
3, 4 |
694 |
|
NR_033328.1 |
|
4, 5 |
741 |
|
NR_033329.1 |
|
4, 5 |
741 |
|
XM_006715753.2 |
XP_006715816.1
|
3, 4 |
605 |
|
XM_006715754.2 |
XP_006715817.1
|
4, 5 |
688 |
|
XM_006715755.2 |
XP_006715818.1
|
4, 5 |
907 |
|
XM_006715756.2 |
XP_006715819.1
|
3, 4 |
776 |
|
XM_006715757.3 |
XP_006715820.1
|
3, 4 |
605 |
|
XM_017012439.1 |
XP_016867928.1
|
4, 5 |
907 |
|
XM_017012440.1 |
XP_016867929.1
|
3, 4 |
605 |
|
XM_017012441.1 |
XP_016867930.1
|
4, 5 |
688 |
|
GenBank mRNA |
AF111113.1 |
- |
3, 4 |
494 |
|
AK056390.1 |
- |
4, 5 |
692 |
|
AK290479.1 |
- |
3, 4 |
404 |
|
AK297595.1 |
- |
2, 3 |
451 |
|
AK299006.1 |
- |
3, 4 |
336 |
|
AK302995.1 |
- |
4, 5 |
464 |
|
AL136597.1 |
- |
3, 4 |
556 |
|
AY028802.1 |
- |
3, 4 |
259 |
|
BC009555.1 |
- |
4, 5 |
657 |
|
BC039585.1 |
- |
3, 4 |
552 |
5527346 |
EF560731.1 |
- |
4, 5 |
460 |
|
View Full Product Details