Gene Transcripts

Gene Symbol
ANKRD46
Entrez Gene ID
157567
Gene Name
ankyrin repeat domain 46
Gene Aliases
ANK-S, GENX-115279
Chromosome Location
Chr. 8: 100509752 - 100559786 on Build GRCh38
UniGene ID
Hs.530199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270377.1 NP_001257306.1 3 291
NM_001270378.1 NP_001257307.1 3 889
NM_001270379.1 NP_001257308.1 3 291
NM_198401.3 NP_940683.1 4 396
XM_006716509.1 XP_006716572.1 3 432
XM_017013106.1 XP_016868595.1 3 354
GenBank mRNA AB077205.1 - 4 394
AK095235.1 - 3 266
AL832842.1 - 3 251
BC050396.1 - 3 247 6065488
BC060843.1 - 3 264 30341860
DA503253.1 - 3 291

View Full Product Details