Gene Transcripts
- Gene Symbol
- STYK1
- Entrez Gene ID
- 55359
- Gene Name
- serine/threonine/tyrosine kinase 1
- Gene Aliases
- NOK, SuRTK106
- Chromosome Location
- Chr. 12: 10618923 - 10674318 on Build GRCh38
- UniGene ID
- Hs.24979
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018423.2 |
NP_060893.2
|
8, 9 |
1448 |
|
XM_005253417.2 |
XP_005253474.1
|
7, 8 |
1348 |
|
XM_011520736.1 |
XP_011519038.1
|
9, 10 |
1251 |
|
XM_011520737.2 |
XP_011519039.1
|
8, 9 |
1288 |
|
XM_011520738.2 |
XP_011519040.1
|
8, 9 |
1443 |
|
XM_017019579.1 |
XP_016875068.1
|
8, 9 |
1231 |
|
GenBank mRNA |
AF251059.1 |
- |
8, 9 |
1448 |
|
AK313906.1 |
- |
8, 9 |
1183 |
|
AL353940.1 |
- |
8, 9 |
1196 |
|
AY563054.1 |
- |
6, 7 |
927 |
|
BC093822.1 |
- |
6, 7 |
949 |
7939667 |
BC093824.1 |
- |
6, 7 |
949 |
7939669 |
KP729000.1 |
- |
6, 7 |
927 |
|
View Full Product Details