Gene Transcripts

Gene Symbol
UFC1
Entrez Gene ID
51506
Gene Name
ubiquitin-fold modifier conjugating enzyme 1
Gene Aliases
HSPC155
Chromosome Location
Chr. 1: 161132754 - 161158856 on Build GRCh38
UniGene ID
Hs.301412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016406.3 NP_057490.2 1 364
XM_005245254.1 XP_005245311.1 1 316
XM_017001450.1 XP_016856939.1 2 209
XM_017001451.1 XP_016856940.1 2 473
GenBank mRNA AB154405.1 - 1 110
AF151884.1 - 1 331
AF161504.1 - 1 350
AK292776.1 - 1 316
BC005187.1 - 1 356 3961569
CF128121.1 - 1 363

View Full Product Details