Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001113546.1 | NP_001107018.1 | 11 | 2896 | |
NM_001113547.1 | NP_001107019.1 | 8 | 2777 | ||
NM_001243775.1 | NP_001230704.1 | 5 | 2504 | ||
NM_016357.4 | NP_057441.1 | 11 | 2893 | ||
XM_011538455.2 | XP_011536757.1 | 11 | 2956 | ||
XM_017019429.1 | XP_016874918.1 | 6 | 2406 | ||
XM_017019430.1 | XP_016874919.1 | 11 | 3247 | ||
GenBank mRNA | AF157325.1 | - | 5 | 2503 | |
AF198454.1 | - | 11 | 2820 | ||
AF198455.1 | - | 8 | 2715 | ||
AK000057.1 | - | 5 | 1819 | ||
AK000335.1 | - | 8 | 2417 | ||
AL136911.1 | - | 11 | 2812 | ||
BC010664.1 | - | 1 | 1270 | ||
BC110815.1 | - | 11 | 2830 | 5115521 |