Gene Transcripts

Gene Symbol
RAB9B
Entrez Gene ID
51209
Gene Name
RAB9B, member RAS oncogene family
Gene Aliases
RAB9L, Rab-9L
Chromosome Location
Chr. X: 103822325 - 103832282 on Build GRCh38
UniGene ID
Hs.522736
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016370.2 NP_057454.1 3 1063
GenBank mRNA AB036693.1 - 3 1009

View Full Product Details