Gene Transcripts
- Gene Symbol
- ZC3HC1
- Entrez Gene ID
- 51530
- Gene Name
- zinc finger C3HC-type containing 1
- Gene Aliases
- HSPC216, NIPA
- Chromosome Location
- Chr. 7: 130018286 - 130051451 on Build GRCh38
- UniGene ID
- Hs.194157
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282190.1 |
NP_001269119.1
|
6, 7 |
745 |
|
NM_001282191.1 |
NP_001269120.1
|
5, 6 |
699 |
|
NM_016478.4 |
NP_057562.3
|
5, 6 |
699 |
|
XM_005250403.2 |
XP_005250460.1
|
3, 4 |
359 |
|
XM_011516288.2 |
XP_011514590.1
|
4, 5 |
525 |
|
XM_011516289.2 |
XP_011514591.1
|
4, 5 |
910 |
|
XM_011516290.2 |
XP_011514592.1
|
3, 4 |
2249 |
|
XM_017012287.1 |
XP_016867776.1
|
5, 6 |
694 |
|
XM_017012288.1 |
XP_016867777.1
|
2, 3 |
1201 |
|
GenBank mRNA |
AF151050.1 |
- |
5, 6 |
621 |
|
AJ537494.1 |
- |
5, 6 |
614 |
|
AK001317.1 |
- |
6, 7 |
668 |
|
AK022373.1 |
- |
5, 6 |
622 |
|
BC000190.1 |
- |
5, 6 |
607 |
|
BC011551.1 |
- |
5, 6 |
615 |
4561164 |
BM013594.1 |
- |
5, 6 |
617 |
|
View Full Product Details