Gene Transcripts
- Gene Symbol
- PPHLN1
- Entrez Gene ID
- 51535
- Gene Name
- periphilin 1
- Gene Aliases
- CR, HSPC206, HSPC232
- Chromosome Location
- Chr. 12: 42326126 - 42448621 on Build GRCh38
- UniGene ID
- Hs.444157
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001143787.1 |
NP_001137259.1
|
5, 6 |
501 |
|
NM_016488.6 |
NP_057572.5
|
6, 7 |
666 |
|
NM_201438.1 |
NP_958846.1
|
6, 7 |
544 |
|
NM_201439.1 |
NP_958847.1
|
6, 7 |
666 |
|
NM_201440.1 |
NP_958848.1
|
6, 7 |
544 |
|
NM_201515.1 |
NP_958923.1
|
7, 8 |
709 |
|
XM_011538459.2 |
XP_011536761.1
|
7, 8 |
687 |
|
XM_011538467.2 |
XP_011536769.1
|
6, 7 |
526 |
|
XM_017019434.1 |
XP_016874923.1
|
7, 8 |
687 |
|
XM_017019435.1 |
XP_016874924.1
|
7, 8 |
686 |
|
XM_017019436.1 |
XP_016874925.1
|
7, 8 |
687 |
|
XM_017019437.1 |
XP_016874926.1
|
6, 7 |
644 |
|
XM_017019438.1 |
XP_016874927.1
|
6, 7 |
644 |
|
XM_017019439.1 |
XP_016874928.1
|
7, 8 |
687 |
|
XM_017019444.1 |
XP_016874933.1
|
6, 7 |
526 |
|
XM_017019445.1 |
XP_016874934.1
|
5, 6 |
520 |
|
XM_017019446.1 |
XP_016874935.1
|
5, 6 |
519 |
|
GenBank mRNA |
AF151066.1 |
- |
6, 7 |
521 |
|
AK055690.1 |
- |
6, 7 |
654 |
|
AK129910.1 |
- |
7, 8 |
709 |
|
AK303612.1 |
- |
5, 6 |
501 |
|
AY039238.1 |
- |
6, 7 |
648 |
|
AY157850.1 |
- |
7, 8 |
705 |
|
BC025306.1 |
- |
5, 6 |
468 |
|
BC039832.1 |
- |
6, 7 |
511 |
6051282 |
BC040449.1 |
- |
6, 7 |
511 |
|
CF242917.1 |
- |
5, 6 |
516 |
|
View Full Product Details