Gene Transcripts
- Gene Symbol
- DNAJC27
- Entrez Gene ID
- 51277
- Gene Name
- DnaJ heat shock protein family (Hsp40) member C27
- Gene Aliases
- RBJ, RabJS
- Chromosome Location
- Chr. 2: 24943636 - 24972094 on Build GRCh38
- UniGene ID
- Hs.434993
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001198559.1 |
NP_001185488.1
|
2, 3 |
369 |
|
NM_016544.2 |
NP_057628.1
|
2, 3 |
369 |
|
XM_011532902.1 |
XP_011531204.1
|
2, 3 |
344 |
|
XM_017004269.1 |
XP_016859758.1
|
2, 3 |
400 |
|
XM_017004270.1 |
XP_016859759.1
|
2, 3 |
260 |
|
GenBank mRNA |
AF178983.1 |
- |
2, 3 |
203 |
|
AK294726.1 |
- |
2, 3 |
153 |
|
AL137731.1 |
- |
2, 3 |
337 |
|
AY094594.1 |
- |
2, 3 |
179 |
|
BC034049.1 |
- |
2, 3 |
230 |
4815297 |
View Full Product Details