Gene Transcripts
- Gene Symbol
- TDP2
- Entrez Gene ID
- 51567
- Gene Name
- tyrosyl-DNA phosphodiesterase 2
- Gene Aliases
- AD022, EAP2, EAPII, TTRAP, dJ30M3.3, hTDP2
- Chromosome Location
- Chr. 6: 24649977 - 24666887 on Build GRCh38
- UniGene ID
- Hs.403010
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016614.2 |
NP_057698.2
|
1, 2 |
200 |
|
GenBank mRNA |
AF201687.1 |
- |
1, 2 |
200 |
|
AF223469.1 |
- |
1, 2 |
191 |
|
AJ269473.1 |
- |
1, 2 |
196 |
|
AK296623.1 |
- |
1, 2 |
110 |
|
AK298699.1 |
- |
1, 2 |
405 |
|
AK308836.1 |
- |
1, 2 |
203 |
|
BC017553.2 |
- |
1, 2 |
212 |
3920790 |
BC110375.1 |
- |
1, 2 |
181 |
5737150 |
JN104587.1 |
- |
1, 2 |
295 |
|
View Full Product Details