Gene Transcripts
- Gene Symbol
- TNFSF8
- Entrez Gene ID
- 944
- Gene Name
- tumor necrosis factor superfamily member 8
- Gene Aliases
- CD153, CD30L, CD30LG, TNLG3A
- Chromosome Location
- Chr. 9: 114893343 - 114930595 on Build GRCh38
- UniGene ID
- Hs.494901
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001244.3 |
NP_001235.1
|
4 |
1165 |
|
GenBank mRNA |
L09753.1 |
- |
4 |
987 |
|
View Full Product Details