Gene Transcripts
- Gene Symbol
- KLHDC4
- Entrez Gene ID
- 54758
- Gene Name
- kelch domain containing 4
- Gene Aliases
- Chromosome Location
- Chr. 16: 87693537 - 87765997 on Build GRCh38
- UniGene ID
- Hs.123450
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001184856.1 |
NP_001171785.1
|
1, 2 |
203 |
|
NM_017566.3 |
NP_060036.2
|
1, 2 |
203 |
|
XM_005255994.3 |
XP_005256051.1
|
1, 2 |
208 |
|
XM_005255999.1 |
XP_005256056.1
|
1, 2 |
202 |
|
XM_006721201.1 |
XP_006721264.1
|
1, 2 |
208 |
|
XM_006721202.1 |
XP_006721265.1
|
1, 2 |
208 |
|
XM_006721203.2 |
XP_006721266.1
|
1, 2 |
208 |
|
XM_006721204.3 |
XP_006721267.1
|
1, 2 |
208 |
|
XM_017023344.1 |
XP_016878833.1
|
1, 2 |
208 |
|
XM_017023345.1 |
XP_016878834.1
|
1, 2 |
143 |
|
GenBank mRNA |
AK023986.1 |
- |
1, 2 |
160 |
|
BC011680.2 |
- |
1, 2 |
187 |
4303424 |
BC022969.1 |
- |
1, 2 |
147 |
5112326 |
BP215584.1 |
- |
1, 2 |
203 |
|
View Full Product Details