Gene Transcripts

Gene Symbol
DIRAS2
Entrez Gene ID
54769
Gene Name
DIRAS family GTPase 2
Gene Aliases
Di-Ras2
Chromosome Location
Chr. 9: 90609832 - 90643105 on Build GRCh38
UniGene ID
Hs.165636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017594.4 NP_060064.2 2 475
GenBank mRNA AB076889.1 - 1 115
AK122986.1 - 2 196
AK124628.1 - 2 170
BC008065.2 - 2 155 3029902
DA221006.1 - 2 475

View Full Product Details