Gene Transcripts
- Gene Symbol
- Hps5
- Entrez Gene ID
- 246694
- Gene Name
- Hermansky-Pudlak syndrome 5 homolog (human)
- Gene Aliases
- AI646796, AL022647, C85120, haze, hz, maroon, mr, ru-2, ru2
- Chromosome Location
- Chr. 7: 46760466 - 46796064 on Build GRCm38
- UniGene ID
- Mm.379442
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001005247.2 |
NP_001005247.2
|
6, 7 |
1115 |
|
NM_001005248.2 |
NP_001005248.2
|
6, 7 |
1091 |
|
NM_001167864.1 |
NP_001161336.1
|
6, 7 |
1115 |
|
GenBank mRNA |
AF534396.1 |
- |
6, 7 |
912 |
|
AF534397.1 |
- |
6, 7 |
888 |
|
AF534398.1 |
- |
4, 5 |
477 |
|
AF534399.1 |
- |
6, 7 |
912 |
|
AK141088.1 |
- |
6, 7 |
913 |
|
AK154612.1 |
- |
6, 7 |
933 |
|
AK157182.1 |
- |
6, 7 |
882 |
|
BC046405.1 |
- |
1, 2 |
94 |
|
BC082542.1 |
- |
6, 7 |
1115 |
6405811 |
View Full Product Details