Gene Transcripts

Gene Symbol
CDAN1
Entrez Gene ID
146059
Gene Name
codanin 1
Gene Aliases
CDA1, CDAI, CDAN1A, DLT, PRO1295
Chromosome Location
Chr. 15: 42723557 - 42737219 on Build GRCh38
UniGene ID
Hs.599232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138477.2 NP_612486.2 8 1463
XM_005254176.4 XP_005254233.1 8 1378
XM_011521270.2 XP_011519572.1 8 1403
XM_011521271.2 XP_011519573.1 8 1400
XM_011521274.2 XP_011519576.1 5 449
GenBank mRNA AF525398.1 - 8 1460
BC052568.1 - 8 1351
BC066640.1 - 8 1351 6577335

View Full Product Details