Gene Transcripts

Gene Symbol
Fastkd1
Entrez Gene ID
320720
Gene Name
FAST kinase domains 1
Gene Aliases
5330408N05Rik, mKIAA1800
Chromosome Location
Chr. 2: 69686820 - 69712606 on Build GRCm38
UniGene ID
Mm.289566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177244.3 NP_796218.2 1 351
GenBank mRNA AK030411.1 - 1242
AK083186.1 - 1262
BC075689.1 - 1 351 30609755

View Full Product Details