Gene Transcripts

Gene Symbol
Tnpo3
Entrez Gene ID
320938
Gene Name
transportin 3
Gene Aliases
5730544L10Rik, C430013M08Rik, C81142, D6Ertd313e, Trn-SR, mKIAA4133
Chromosome Location
Chr. 6: 29540827 - 29610110 on Build GRCm38
UniGene ID
Mm.276381
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177296.4 NP_796270.2 19 2443
GenBank mRNA AK077714.1 - 3 273
AK220184.1 - 12 1346
BC023273.1 - 7 628
BC052756.1 - 8 764
BC064646.1 - 19 2443 6313452
BC075678.1 - 19 2656 30619729

View Full Product Details