Gene Transcripts

Gene Symbol
Mrfap1
Entrez Gene ID
282585
Gene Name
Morf4 family associated protein 1
Gene Aliases
Pgr1, RGD1306635
Chromosome Location
Chr. 14: 78973384 - 78975161 on Build Rnor_6.0
UniGene ID
Rn.3393
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009264.1 NP_001009264.1 2 526
GenBank mRNA AF526268.1 - 3 476
BC088308.1 - 2 526 7324285
FQ228750.1 - 2 546

View Full Product Details