Gene Transcripts

Gene Symbol
Tmco1
Entrez Gene ID
289196
Gene Name
transmembrane and coiled-coil domains 1
Gene Aliases
Chromosome Location
Chr. 13: 85465007 - 85488851 on Build Rnor_6.0
UniGene ID
Rn.89308
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009631.1 NP_001009631.1 436
GenBank mRNA AF531432.1 - 359
BC088319.1 - 436 7325542

View Full Product Details