Gene Transcripts

Gene Symbol
DEF8
Entrez Gene ID
54849
Gene Name
differentially expressed in FDCP 8 homolog (mouse)
Gene Aliases
Chromosome Location
Chr. 16: 89948697 - 89968060 on Build GRCh38
UniGene ID
Hs.62771
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242816.1 NP_001229745.1 5 439
NM_001242817.1 NP_001229746.1 4 335
NM_001242818.1 NP_001229747.1 5 469
NM_001242819.1 NP_001229748.1 4 382
NM_001242820.1 NP_001229749.1 6 709
NM_001242821.1 NP_001229750.1 4 372
NM_001242822.1 NP_001229751.1 5 466
NM_017702.3 NP_060172.1 5 469
NM_207514.2 NP_997397.1 5 594
XM_005256318.2 XP_005256375.2 5 445
XM_011523156.2 XP_011521458.1 5 445
XM_011523157.2 XP_011521459.1 5 445
XM_011523158.1 XP_011521460.1 5 445
XM_011523159.2 XP_011521461.1 5 393
XM_011523160.2 XP_011521462.1 5 546
XM_011523161.2 XP_011521463.1 5 527
XM_011523162.2 XP_011521464.1 4 406
XM_017023358.1 XP_016878847.1 5 445
XM_017023359.1 XP_016878848.1 5 445
XM_017023360.1 XP_016878849.1 5 445
XM_017023361.1 XP_016878850.1 5 393
XM_017023362.1 XP_016878851.1 5 393
XM_017023363.1 XP_016878852.1 5 527
XM_017023364.1 XP_016878853.1 5 393
XM_017023365.1 XP_016878854.1 5 546
XM_017023366.1 XP_016878855.1 4 1510
XM_017023367.1 XP_016878856.1 4 406
XM_017023368.1 XP_016878857.1 4 1510
XM_017023369.1 XP_016878858.1 4 406
GenBank mRNA AK000193.1 - 5 405
AK095051.1 - 5 550
AK131370.1 - 5 375
AK225539.1 - 5 410
AK295880.1 - 6 671
AK296410.1 - 4 271
AK303510.1 - 4 316
BC015482.2 - 4 392
BC105592.1 - 4 312
BC130482.1 - 4 355
BC144038.1 - 4 355
BM712309.1 - 4 375
DC332872.1 - 5 469

View Full Product Details