Gene Transcripts

Gene Symbol
Nubp2
Entrez Gene ID
287125
Gene Name
nucleotide binding protein 2
Gene Aliases
Chromosome Location
Chr. 10: 14244201 - 14247886 on Build Rnor_6.0
UniGene ID
Rn.203395
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011891.1 NP_001011891.1 7 1365
XM_008767558.1 XP_008765780.1 7 1378
GenBank mRNA BC079386.1 - 7 1365

View Full Product Details