Gene Transcripts

Gene Symbol
DOCK10
Entrez Gene ID
55619
Gene Name
dedicator of cytokinesis 10
Gene Aliases
DRIP2, Nbla10300, ZIZ3
Chromosome Location
Chr. 2: 224765090 - 225042689 on Build GRCh38
UniGene ID
Hs.46578
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290263.1 NP_001277192.1 45 5203
NM_014689.2 NP_055504.2 45 5346
XM_005246673.4 XP_005246730.1 45 5205
XM_006712617.3 XP_006712680.1 45 5370
XM_017004441.1 XP_016859930.1 45 5373
XM_017004442.1 XP_016859931.1 45 5349
XM_017004443.1 XP_016859932.1 45 5422
XM_017004444.1 XP_016859933.1 44 5329
XM_017004445.1 XP_016859934.1 45 5203
XM_017004446.1 XP_016859935.1 44 5227
XM_017004447.1 XP_016859936.1 45 5345
GenBank mRNA AF250396.1 - 5 715
AK000227.1 - 3 315
AK293847.1 - 11 1272
AK293879.1 - 16 1752
AK307914.1 - 5 707
BC015018.1 - 3 313
CR749492.1 - 20 2191
EU236710.2 - 45 5200

View Full Product Details