Gene Transcripts

Gene Symbol
Hbs1l
Entrez Gene ID
293408
Gene Name
HBS1-like translational GTPase
Gene Aliases
Chromosome Location
Chr. 1: 16819170 - 16896221 on Build Rnor_6.0
UniGene ID
Rn.98198
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011934.1 NP_001011934.1 4, 5 490
GenBank mRNA BC079463.1 - 4, 5 490

View Full Product Details