Gene Transcripts

Gene Symbol
Sept1
Entrez Gene ID
293507
Gene Name
septin 1
Gene Aliases
Chromosome Location
Chr. 1: 198658744 - 198662610 on Build Rnor_6.0
UniGene ID
Rn.68675
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012460.1 NP_001012478.1 11 1360
GenBank mRNA BC089897.1 - 11 1360

View Full Product Details