Gene Transcripts

Gene Symbol
MTMR8
Entrez Gene ID
55613
Gene Name
myotubularin related protein 8
Gene Aliases
Chromosome Location
Chr. X: 64268081 - 64395453 on Build GRCh38
UniGene ID
Hs.442892
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017677.3 NP_060147.2 4 466
GenBank mRNA AK000133.1 - 2 125
AK225231.1 - 2 125
BC012399.1 - 4 444 3864128
DA691850.1 - 4 466

View Full Product Details