Gene Transcripts

Gene Symbol
Lca5
Entrez Gene ID
300866
Gene Name
Leber congenital amaurosis 5
Gene Aliases
RGD1308555
Chromosome Location
Chr. 8: 90926309 - 90984284 on Build Rnor_6.0
UniGene ID
Rn.208333
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013954.1 NP_001013976.1 9 2304
XM_008766249.1 XP_008764471.1 8 2248
XM_008766250.1 XP_008764472.1 7 2129
XM_008766251.1 XP_008764473.1 9 2339
XM_008766252.1 XP_008764474.1 8 2192
XM_008766253.1 XP_008764475.1 8 2635
XM_008766255.1 XP_008764477.1 9 2364
XM_008766256.1 XP_008764478.1 8 2170
XM_008766257.1 XP_008764479.1 8 2809
XM_008766258.1 XP_008764480.1 8 5833
GenBank mRNA BC085805.1 - 9 2304

View Full Product Details