Gene Transcripts

Gene Symbol
Slc46a1
Entrez Gene ID
303333
Gene Name
solute carrier family 46 (folate transporter), member 1
Gene Aliases
RGD1309472
Chromosome Location
Chr. 10: 65728484 - 65741708 on Build Rnor_6.0
UniGene ID
Rn.153975
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013969.1 NP_001013991.1 5 1549
XM_006246939.2 XP_006247001.1 6 1857
GenBank mRNA BC089868.1 - 5 1549
FQ231269.1 - 5 1575

View Full Product Details